(2-Fluoro-5-nitro-3-(trifluoromethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A422784-1G A422784-250MG A422784-5G
77143-174EA 156.56 USD
77143-174 77143-176 77143-178
(2-Fluoro-5-nitro-3-(trifluoromethyl)phenyl)boronic acid 97%
(2-Fluoro-5-nitro-3-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₄BF₄NO₄
MDL Number: MFCD20529438
CAS Number: 2096335-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR