Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I) 97%

Supplier: Ambeed
Warning

A422154-1G A422154-250MG
77143-170EA 52.53 USD
77143-170 77143-172
Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I) 97%
Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I)

Precaución: for Research Use Only

Formula: C₂₁H₂₄ClCuN₂
MDL Number: MFCD15144833
CAS Number: 873779-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR