Benzhydrylamine 98%

Supplier: Ambeed
Warning

Synonyms: α-Phenylbenzylamine, α-Aminodiphenylmethane, Diphenylmethanamine, Aminodiphenylmethane, 1,1-Diphenylmethanamine, Benzhydrylamin

A421685-100G A421685-10G A421685-25G A421685-500G A421685-1000G
77143-156EA 45.33 USD
77143-156 77143-158 77143-160 77143-162 77616-828
Benzhydrylamine 98%
Benzhydrylamine

Precaución: for Research Use Only

Formula: C₁₃H₁₃N
MDL Number: MFCD00008059
CAS Number: 91-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR