(2-Bromo-4-isopropylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-isopropylphebylboronic acid

A420826-100MG A420826-1G A420826-250MG
77143-150EA 36.08 USD
77143-150 77143-152 77143-154
(2-Bromo-4-isopropylphenyl)boronic acid 97%
(2-Bromo-4-isopropylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BBrO₂
MW: 242.90 g/mol
MDL Number: MFCD18434420
CAS Number: 2096338-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR