6-Aminohexane-1-thiol hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A420028-100MG
A420028-250MG
A420028-1G
77143-142EA
18.54
USD
77143-142
77143-144
77551-268
6-Aminohexane-1-thiol hydrochloride 95%
6-Aminohexane-1-thiol hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₆ClNS |
MDL Number:
MFCD18910700 CAS Number: 31098-40-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...