(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-Triphenylethylene Glycol, (S)-1,1,2-Triphenyl-1,2-ethanediol

A419285-1G A419285-250MG A419285-5G
77143-130EA 23.68 USD
77143-130 77143-132 77143-134
(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol 97%
(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₂
MDL Number: MFCD00075492
CAS Number: 108998-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR