(4-Cyanophenoxy)acetic acid 96%

Supplier: Ambeed
Warning

A418950-250MG A418950-5G A418950-1G
77143-126EA 15.45 USD
77143-126 77143-128 77572-308
(4-Cyanophenoxy)acetic acid 96%
(4-Cyanophenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MDL Number: MFCD01250758
CAS Number: 1878-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR