3-Bromo-6-fluoro-2-methylaniline 97%

Supplier: Ambeed
Warning

A418552-100MG A418552-1G A418552-250MG A418552-5G
77143-118EA 25.75 USD
77143-118 77143-120 77143-122 77143-124
3-Bromo-6-fluoro-2-methylaniline 97%
3-Bromo-6-fluoro-2-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MDL Number: MFCD23704130
CAS Number: 1227210-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR