(2-Chloro-4-methylphenyl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-methylbenzyl alcohol

A417461-10G A417461-1G A417461-5G
77143-112EA 615.33 USD
77143-112 77143-114 77143-116
(2-Chloro-4-methylphenyl)methanol 97%
(2-Chloro-4-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.60 g/mol
MDL Number: MFCD18393674
CAS Number: 39652-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR