(R)-1-[α-(4-Chlorophenyl)benzyl]piperazine 95%

Supplier: Ambeed
Warning

Synonyms: Chlorobenzhydryl Piperazine, (R)-1-((4-Chlorophenyl)(phenyl)methyl)piperazine

A416464-100G A416464-10G A416464-1G A416464-250MG A416464-5G A416464-25G
77143-102EA 629.34 USD
77143-102 77143-104 77143-106 77143-108 77143-110 77686-148
(R)-1-[α-(4-Chlorophenyl)benzyl]piperazine 95%
(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine

Precaución: for Research Use Only

Formula: C₁₇H₁₉ClN₂
MDL Number: MFCD11519277
CAS Number: 300543-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR