Pyridine-2,3-dicarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Quinolinic acid

A415699-10G A415699-25G A415699-500G A415699-100G
77143-084EA 5.31 USD
77143-084 77143-086 77143-088 77594-284
Pyridine-2,3-dicarboxylic acid 98%
Pyridine-2,3-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₄
MW: 167.12 g/mol
MDL Number: MFCD00006295
CAS Number: 89-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR