4-Amino-3-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-nitrobenzoic Acid

A414128-100G A414128-10G A414128-25G A414128-500G
77143-076EA 38.2 USD
77143-076 77143-078 77143-080 77143-082
4-Amino-3-nitrobenzoic acid 95%
4-Amino-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
MDL Number: MFCD00017009
CAS Number: 1588-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR