(3-Cyano-4-hydroxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A414113-1G A414113-250MG
77143-072EA 154.51 USD
77143-072 77143-074
(3-Cyano-4-hydroxyphenyl)boronic acid 97%
(3-Cyano-4-hydroxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BNO₃
MDL Number: MFCD18394168
CAS Number: 1637208-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR