N-(9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(1-hydroxy-2,2-bis(4-methoxyphenyl)-2-phenylethyl)tetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-1H-purin-2-yl)isobutyramide 95%

Supplier: Ambeed
Warning

Synonyms: 5'-O-DMT-ibu-rG

A412698-1G A412698-5G
77143-070EA 40.18 USD
77143-070 77616-796
N-(9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(1-hydroxy-2,2-bis(4-methoxyphenyl)-2-phenylethyl)tetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-1H-purin-2-yl)isobutyramide 95%
N-(9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(1-hydroxy-2,2-bis(4-methoxyphenyl)-2-phenylethyl)tetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-1H-purin-2-yl)isobutyramide

Precaución: for Research Use Only

Formula: C₃₅H₃₇N₅O₈
MW: 655.69 g/mol
MDL Number: MFCD00797504
CAS Number: 81246-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR