(-)-Ambroxide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ambroxide, (3aR,5aS,9aS,9bR)-3a,6,6,9a-Tetramethyldodecahydronaphtho[2,1-b]furan, 1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.0(4,9)]tridecane
A412557-1G
A412557-25G
A412557-25MG
A412557-5G
A412557-5MG
77143-064EA
8.49
USD
77143-064
77143-066
77143-068
77686-141
77686-142
(-)-Ambroxide 97%
(-)-Ambroxide
Precaución: for Research Use Only
Formula:
C₁₆H₂₈O MW: 236.40 g/mol |
MDL Number:
MFCD00134491 CAS Number: 6790-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...