1-Methyl-4-nitro-1H-pyrazole-5-carbonitrile 97%

Supplier: Ambeed
Warning

A412516-100MG A412516-250MG A412516-5G
77143-058EA 33.95 USD
77143-058 77143-060 77143-062
1-Methyl-4-nitro-1H-pyrazole-5-carbonitrile 97%
1-Methyl-4-nitro-1H-pyrazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄N₄O₂
MW: 152.11 g/mol
MDL Number: MFCD00463995
CAS Number: 42110-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR