(4-((Ethoxymethyl)thio)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A411203-1G A411203-250MG A411203-5G
77143-052EA 52.53 USD
77143-052 77143-054 77143-056
(4-((Ethoxymethyl)thio)phenyl)boronic acid 97%
(4-((Ethoxymethyl)thio)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃S
MDL Number: MFCD19237160
CAS Number: 2096331-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR