(8-Quinolinolato)lithium 98%

Supplier: Ambeed
Warning

Synonyms: Lithium quinolin-8-olate, Liq

A411195-1G A411195-250MG A411195-5G
77143-046EA 7.21 USD
77143-046 77143-048 77143-050
(8-Quinolinolato)lithium 98%
(8-Quinolinolato)lithium

Precaución: for Research Use Only

Formula: C₉H₆LiNO
MW: 151.09 g/mol
MDL Number: MFCD00152778
CAS Number: 25387-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR