2,4,6-Trifluorobenzamide 98%

Supplier: Ambeed
Warning

A410659-1G A410659-25G A410659-5G
77143-040EA 19.57 USD
77143-040 77143-042 77143-044
2,4,6-Trifluorobenzamide 98%
2,4,6-Trifluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₄F₃NO
MDL Number: MFCD00061199
CAS Number: 82019-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR