2-(Methylsulfinyl)pyrimidine 95%

Supplier: Ambeed
Warning

A410272-100MG A410272-1G A410272-250MG
77143-034EA 25.76 USD
77143-034 77143-036 77143-038
2-(Methylsulfinyl)pyrimidine 95%
2-(Methylsulfinyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₆N₂OS
MW: 142.18 g/mol
MDL Number: MFCD11046328
CAS Number: 14080-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR