N-Methyl 4-boronobenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methylsulphamoylbenzeneboronic acid, (4-(N-Methylsulphamoyl)phenyl)boronic acid

A406831-1G A406831-250MG A406831-5G
77143-000EA 133.68 USD
77143-000 77143-002 77143-004
N-Methyl 4-boronobenzenesulfonamide 98%
N-Methyl 4-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₄S
MW: 215.03 g/mol
MDL Number: MFCD06659864
CAS Number: 226396-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR