m-PEG4-amine 96%

Supplier: Ambeed
Danger

Synonyms: 3,6,9,12-Tetraoxatridecanamine

A405944-250MG A405944-1G A405944-5G A405944-25G
77142-988EA 53.05 USD
77142-988 77551-218 77551-220 77686-117
m-PEG4-amine 96%
m-PEG4-amine

Precaución: for Research Use Only

Formula: C₉H₂₁NO₄
MW: 207.27 g/mol
MDL Number: MFCD11041099
CAS Number: 85030-56-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR