2-((3-Chlorophenyl)amino)acetic acid 95+%

Supplier: Ambeed
Warning

A404501-1G A404501-250MG A404501-5G
77142-950EA 32.96 USD
77142-950 77142-952 77142-954
2-((3-Chlorophenyl)amino)acetic acid 95+%
2-((3-Chlorophenyl)amino)acetic acid

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MDL Number: MFCD02255622
CAS Number: 10242-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR