1,4-Anhydroerythritol 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Anhydroerythritol, cis-Tetrahydrofuran-3,4-diol, cis-3,4-Tetrahydrofurandiol

A404396-25G A404396-5G A404396-100G A404396-1G
77142-946EA 131.84 USD
77142-946 77142-948 77616-746 77616-748
1,4-Anhydroerythritol 98%
1,4-Anhydroerythritol

Precaución: for Research Use Only

Formula: C₄H₈O₃
MW: 104.11 g/mol
MDL Number: MFCD00075210
CAS Number: 4358-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR