(2-Methyl-1,3-thiazol-4-yl)methanol 96%

Supplier: Ambeed
Danger

Synonyms: (2-Methylthiazol-4-yl)methanol

A403592-1G A403592-250MG A403592-5G
77142-940EA 38.2 USD
77142-940 77142-942 77142-944
(2-Methyl-1,3-thiazol-4-yl)methanol 96%
(2-Methyl-1,3-thiazol-4-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₇NOS
MW: 129.1802 g/mol
MDL Number: MFCD08703637
CAS Number: 76632-23-0
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR