2-Amino-4-bromo-5-chloro-3-fluorobenzoic acid 95%

Supplier: Ambeed
Warning

A403194-1G A403194-5G
77142-938EA 105.03 USD
77142-938 77568-814
2-Amino-4-bromo-5-chloro-3-fluorobenzoic acid 95%
2-Amino-4-bromo-5-chloro-3-fluorobenzoic acid

Precaución: For research use only.

Formula: C₇H₄BrClFNO₂
MW: 268.47 g/mol
MDL Number: MFCD29058767
CAS Number: 1698027-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR