4-(2-Bromopyrimidin-4-yl)morpholine 95%

Supplier: Ambeed
Warning

A402601-100MG A402601-1G A402601-250MG
77142-932EA 70.02 USD
77142-932 77142-934 77142-936
4-(2-Bromopyrimidin-4-yl)morpholine 95%
4-(2-Bromopyrimidin-4-yl)morpholine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN₃O
MW: 244.08 g/mol
MDL Number: MFCD14702752
CAS Number: 1209459-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR