Benzyl ((2S,3R)-3-hydroxy-1-oxo-1-(((tetrahydro-2H-pyran-2-yl)oxy)amino)butan-2-yl)carbamate 97%

Supplier: Ambeed
Warning

A402578-1G A402578-250MG
77142-926EA 14.86 USD
77142-926 77142-928 77142-930
Benzyl ((2S,3R)-3-hydroxy-1-oxo-1-(((tetrahydro-2H-pyran-2-yl)oxy)amino)butan-2-yl)carbamate 97%
Benzyl ((2S,3R)-3-hydroxy-1-oxo-1-(((tetrahydro-2H-pyran-2-yl)oxy)amino)butan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₇H₂₄N₂O₆
MW: 352.38 g/mol
MDL Number: MFCD27992080
CAS Number: 1820598-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR