5-Nitrobenzimidazole 98%

Supplier: Ambeed
Warning

A401617-100G A401617-25G A401617-500G A401617-5G
77142-918EA 39.14 USD
77142-918 77142-920 77142-922 77142-924
5-Nitrobenzimidazole 98%
5-Nitrobenzimidazole

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MDL Number: MFCD00005604
CAS Number: 94-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR