Methyl 2-aminocyclohexanecarboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A400728-100MG A400728-250MG
77142-902EA 69.01 USD
77142-902 77142-904
Methyl 2-aminocyclohexanecarboxylate hydrochloride 98%
Methyl 2-aminocyclohexanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD13195002
CAS Number: 99419-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR