4-(4-Iodo-1H-pyrazol-1-yl)cyclohexanone 97%

Supplier: Ambeed
Danger

A399235-100MG A399235-1G A399235-250MG A399235-5G
77142-888EA 65.78 USD
77142-888 77142-890 77142-892 77551-200
4-(4-Iodo-1H-pyrazol-1-yl)cyclohexanone 97%
4-(4-Iodo-1H-pyrazol-1-yl)cyclohexanone

Precaución: for Research Use Only

Formula: C₉H₁₁IN₂O
MW: 290.10 g/mol
MDL Number: MFCD29054490
CAS Number: 1227611-94-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR