2-Methyl-5-(4-methylphenyl)pyrazol-3-amine 98%

Supplier: Ambeed
Danger

Synonyms: 1-Methyl-3-(p-tolyl)-1H-pyrazol-5-amine

A398782-10G A398782-250MG A398782-25G A398782-5G
77142-878EA 467.86 USD
77142-878 77142-880 77142-882 77142-884
2-Methyl-5-(4-methylphenyl)pyrazol-3-amine 98%
2-Methyl-5-(4-methylphenyl)pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃
MW: 187.241 g/mol
MDL Number: MFCD06637391
CAS Number: 126417-86-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR