1-Ethyl-2,3-dimethylimidazolium bromide 99+%

Supplier: Ambeed
Warning

A397855-100G A397855-25G A397855-5G
77142-870EA 47.38 USD
77142-870 77142-872 77142-874
1-Ethyl-2,3-dimethylimidazolium bromide 99+%
1-Ethyl-2,3-dimethylimidazolium bromide

Precaución: for Research Use Only

Formula: C₇H₁₃BrN₂
MDL Number: MFCD03427614
CAS Number: 98892-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR