5-Bromo-1-methyl-1H-pyrazolo[3,4-c]pyridine 98%
Supplier: Ambeed
|
Warning
|
A397502-100MG
A397502-250MG
A397502-1G
77142-854EA
38.2
USD
77142-854
77142-856
77686-100
5-Bromo-1-methyl-1H-pyrazolo[3,4-c]pyridine 98%
5-Bromo-1-methyl-1H-pyrazolo[3,4-c]pyridine
Precaución: for Research Use Only
Formula:
C₇H₆BrN₃ MW: 212.047 g/mol |
MDL Number:
MFCD22380195 CAS Number: 1337880-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...