4,5-Dimethoxy-2-nitrophenylacetonitrile 97%

Supplier: Ambeed
Danger

A397162-25G A397162-5G
77142-844EA 81.37 USD
77142-844 77142-846
4,5-Dimethoxy-2-nitrophenylacetonitrile 97%
4,5-Dimethoxy-2-nitrophenylacetonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₄
MDL Number: MFCD00033967
CAS Number: 17354-04-0
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR