6-Chloro-3-fluoro-2-methoxypyridine 97%

Supplier: Ambeed
Warning

A397156-1G A397156-250MG
77142-840EA 564.43 USD
77142-840 77142-842
6-Chloro-3-fluoro-2-methoxypyridine 97%
6-Chloro-3-fluoro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFNO
MDL Number: MFCD18415687
CAS Number: 1261473-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR