2-(4-Bromo-2-(trifluoromethoxy)phenyl)acetic acid 99%

Supplier: Ambeed
Warning

A395944-1G A395944-250MG A395944-5G A395944-25G
77142-828EA 29.71 USD
77142-828 77142-830 77142-832 77594-212
2-(4-Bromo-2-(trifluoromethoxy)phenyl)acetic acid 99%
2-(4-Bromo-2-(trifluoromethoxy)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₃
MW: 299.04 g/mol
MDL Number: MFCD12761595
CAS Number: 509142-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR