(4-((tert-Butyldimethylsilyl)oxy)-2,6-difluorophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A395266-100MG A395266-1G A395266-250MG
77142-816EA 31.83 USD
77142-816 77142-818 77142-820
(4-((tert-Butyldimethylsilyl)oxy)-2,6-difluorophenyl)boronic acid 98%
(4-((tert-Butyldimethylsilyl)oxy)-2,6-difluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₉BF₂O₃Si
MW: 288.17 g/mol
MDL Number: MFCD19237185
CAS Number: 2096341-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR