Methyl 2,4-diamino-3-fluoro-5-nitrobenzoate 97%

Supplier: Ambeed
Warning

A395079-100MG A395079-10G A395079-1G A395079-250MG A395079-25G A395079-5G
77142-798EA 6.37 USD
77142-798 77142-800 77142-802 77142-804 77142-806 77142-808
Methyl 2,4-diamino-3-fluoro-5-nitrobenzoate 97%
Methyl 2,4-diamino-3-fluoro-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₈FN₃O₄
MW: 229.16 g/mol
MDL Number: MFCD29920456
CAS Number: 918321-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR