4-Methyl-1H-indole-6-carboxylic acid 97%

Supplier: Ambeed
Warning

A394948-100MG A394948-250MG
77142-794EA 87 USD
77142-794 77142-796
4-Methyl-1H-indole-6-carboxylic acid 97%
4-Methyl-1H-indole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.18 g/mol
MDL Number: MFCD29921869
CAS Number: 1545472-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR