Ethyl-3-(2-furyl)-1H-pyrazole-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(furan-2-yl)-1H-pyrazole-5-carboxylate

A394365-100MG A394365-1G A394365-250MG
77142-786EA 32.89 USD
77142-786 77142-788 77142-790
Ethyl-3-(2-furyl)-1H-pyrazole-5-carboxylate 95%
Ethyl-3-(2-furyl)-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₃
MW: 206.20 g/mol
MDL Number: MFCD09953454
CAS Number: 33545-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR