XL765 98+%

Supplier: Ambeed
Warning

A393898-10MG A393898-1MG A393898-2MG A393898-5MG
77142-770EA 50.93 USD
77142-770 77142-772 77142-774 77142-776
XL765 98+%
XL765

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₆O
MDL Number: MFCD20276656
CAS Number: 934493-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR