SBI-0206965 97%

Supplier: Ambeed
Warning

A391178-10MG A391178-1MG A391178-25MG A391178-2MG A391178-5MG
77142-746EA 53.56 USD
77142-746 77142-748 77142-750 77142-752 77142-754
SBI-0206965 97%
SBI-0206965

Precaución: for Research Use Only

Formula: C₂₁H₂₁BrN₄O₅
MDL Number: MFCD29472244
CAS Number: 1884220-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR