Pyridine-3,4-diyldimethanol hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A390733-1G
A390733-250MG
A390733-5G
77142-730EA
51.99
USD
77142-730
77142-732
77142-734
Pyridine-3,4-diyldimethanol hydrochloride 98%
Pyridine-3,4-diyldimethanol hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₀ClNO₂ MW: 175.61 g/mol |
MDL Number:
MFCD29918533 CAS Number: 53654-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...