Diphenyl(quinuclidin-4-yl)methanol 98%

Supplier: Ambeed
Warning

A390212-1G A390212-250MG A390212-5G
77142-716EA 173.99 USD
77142-716 77142-718 77142-720
Diphenyl(quinuclidin-4-yl)methanol 98%
Diphenyl(quinuclidin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₂₀H₂₃NO
MW: 293.4 g/mol
MDL Number: MFCD22665854
CAS Number: 461648-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR