3,4-Dichloro-6-nitrotoluene 95%

Supplier: Ambeed
Warning

A388069-1G A388069-25G A388069-5G
77142-694EA 6.18 USD
77142-694 77142-696 77142-698
3,4-Dichloro-6-nitrotoluene 95%
3,4-Dichloro-6-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MDL Number: MFCD01109847
CAS Number: 7494-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR