3-Bromo-4-methyl-5-nitropyridin-2-amine 98%

Supplier: Ambeed
Warning

A388016-1G A388016-5G
77142-690EA 80.63 USD
77142-690 77142-692
3-Bromo-4-methyl-5-nitropyridin-2-amine 98%
3-Bromo-4-methyl-5-nitropyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₆BrN₃O₂
MW: 232.03 g/mol
MDL Number: MFCD03427659
CAS Number: 929976-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR