(R)-(-)-3-Chloromandelic acid 95%

Supplier: Ambeed
Warning

Synonyms: (R)-2-(3-Chlorophenyl)-2-hydroxyacetic acid

A387731-1G A387731-250MG A387731-5G
77142-684EA 37.14 USD
77142-684 77142-686 77142-688
(R)-(-)-3-Chloromandelic acid 95%
(R)-(-)-3-Chloromandelic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.59 g/mol
MDL Number: MFCD04038807
CAS Number: 61008-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR