Ethyl 6-chloro-1H-pyrrolo[3,2-c]pyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

A387050-1G A387050-250MG
77142-674EA 197.33 USD
77142-674 77142-676
Ethyl 6-chloro-1H-pyrrolo[3,2-c]pyridine-3-carboxylate 97%
Ethyl 6-chloro-1H-pyrrolo[3,2-c]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MW: 224.64 g/mol
MDL Number: MFCD27922293
CAS Number: 1951441-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR