2-Bromo-6-chloro-4-fluorobenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A386761-100MG
A386761-1G
A386761-250MG
A386761-5G
77142-662EA
38.2
USD
77142-662
77142-664
77142-666
77746-776
2-Bromo-6-chloro-4-fluorobenzaldehyde 97%
2-Bromo-6-chloro-4-fluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₃BrClFO MW: 237.454 g/mol |
MDL Number:
MFCD28733865 CAS Number: 1433990-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...